Schwefel ≥99%, Blume
Glass bottle for solids, 2.500 g
- :7704-34-9
- :S
- :MFCD00085316
- :32.07 g/mol
- :13,09059
- :445 °C (1013 hPa)
- :113…119 °C
- :2,36 g/cm³ (20 °C)
- :168 °C
- :Raumtemperatur
- :231-722-6
- :1350
- :4.1,III
- Packung:Glass bottle for solids
- VE:2.500 g