AuftragsverfolgungKontakt
Grid Storage Box | Avantor
Grid Storage Box
Grid storage box
Artikelnr. SIALG6276-1EA
Anbieter:   Merck
Grid Storage Box
Artikelnr. SIALG6276-1EA
Anbieter:  Merck
Grid Storage Box
Grid storage box
Preis
48,30 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
This storage box is made up of plastic.
  • Anti-static
  • Record card is used to record identity of samples
  • Device is anti-static to prevent handling problems due to repulsion or attraction
Spezifikationen
  • Bezeichnung:
    Grid storage box
  • Seq:
    01