Express Eingabe
Austria
AuftragsverfolgungKontakt
Cyclohexan ≥99% ACS, Riedel-de Haën™

Cyclohexan ≥99% ACS, Riedel-de Haën™

Glasflasche, 6 x 1 l, 6 L
Artikelnr. 179191-6X1L
Entzündlich
Gesundheit
Reizend
Anbieter:   Honeywell Chemicals
Cyclohexan ≥99% ACS, Riedel-de Haën™
Artikelnr. 179191-6X1L
CAS Nummer:  110-82-7
Cyclohexan ≥99% ACS, Riedel-de Haën™
Cyclohexan ≥99% ACS, Riedel-de Haën™

Glasflasche, 6 x 1 l, 6 L

CAS Nummer:  110-82-7
Preis
352,00 €
Einloggen für Verfügbarkeit
1 * 6 L
Product Details & Documents

ACS GENERAL USE SOLVENTS

  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504

Synonyme:  Hexahydrobenzene, Hexamethylene
  • CAS Nummer:110-82-7
  • Formel:C₆H₁₂
  • MDL-Nummer:MFCD00003814
  • Molekulargewicht:84.16 g/mol
  • Merck-Index:13,02752
  • Siedepunkt:81 °C (1,013 hPa)
  • Schmelzpunkt:6,4 °C
  • Dichte:0,78 g/cm³ (20 °C)
  • Blitzpunkt:–18 °C
  • Blitzpunkt:-18 °C
  • Lagertemperatur:Raumtemperatur
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Spezifikationen

Spezifikation Testergebnisse

  • Vapor Pressure:
    168.8 mmHg ( 37.7 °C)
  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.0 %
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Reaction against H₂SO₄:
    complying
  • UV-absorption:
    complying
  • APHA:
    Max. 10
  • Refractive Index:
    n20/D 1.426 (lit.)

Spezifikationen

  • Packung:
    Glasflasche
  • VE:
    6 L
  • Pk Info:
    6 x 1 l

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.