Cyclohexan ≥99% ACS, Riedel-de Haën™
Glasflasche, 1 L
ACS GENERAL USE SOLVENTS
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested to meet ACS general use specifications
Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504
: Hexahydrobenzene, Hexamethylene
- :110-82-7
- :C₆H₁₂
- :MFCD00003814
- :84.16 g/mol
- :13,02752
- :81 °C (1,013 hPa)
- :6,4 °C
- :0,78 g/cm³ (20 °C)
- :–18 °C
- :-18 °C
- :Raumtemperatur
- :203-806-2
- :1145
- :3,II
- Vapor Pressure:168.8 mmHg ( 37.7 °C)
- IR spectrum:complying
- Assay (GC):Min. 99.0 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.02 %
- Reaction against H₂SO₄:complying
- UV-absorption:complying
- APHA:Max. 10
- Refractive Index:n20/D 1.426 (lit.)
- Packung:Glasflasche
- VE:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



