Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Wash buffer, Pierce™

Pierce™ modified Dulbecco's PBS Tween™ 20 buffer, 20X, 7,4, 0,5 L
Artikelnr. PIER28346
Wash buffer, Pierce™
Artikelnr. PIER28346
CAS Nummer:  
Wash buffer, Pierce™
Wash buffer, Pierce™

Pierce™ modified Dulbecco's PBS Tween™ 20 buffer, 20X, 7,4, 0,5 L

CAS Nummer:  
Preis
148,00 €
Einloggen für Verfügbarkeit
1 * 0,5 L
Product Details & Documents

These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.

  • Ready to use without having to reconstitute with ultrapure water
  • Eliminates the possibility of powder remaining in a packet
  • 10X or 20X concentration eliminates time spent waiting for powder to dissolve
  • Storage as concentrated stock minimizes bench space needed for large volume solutions
Spezifikationen

Spezifikationen

  • VE:
    0,5 L
  • Bezeichnung:
    Pierce™ modified Dulbecco's PBS Tween™ 20 buffer, 20X
  • pH-Wert:
    7,4

Spezifikationen