Biopsy specimen processing, CellSafe™
CellSafe™ biopsy capsule
720-2334
: CELLPATH
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
- No tissue damage
- Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
- Bezeichnung:CellSafe™ biopsy capsule