AuftragsverfolgungKontakt
Biopsy specimen processing, CellSafe™ | Avantor
Biopsy specimen processing, CellSafe™
Curettage cassette with stainless steel insert and lid
Artikelnr. 720-2332
Anbieter:   CELLPATH
Biopsy specimen processing, CellSafe™
Artikelnr. 720-2332
Anbieter:  CELLPATH
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Curettage cassette with stainless steel insert and lid
Preis
356,00 €
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Spezifikationen
  • Bezeichnung:
    Curettage cassette with stainless steel insert and lid