AuftragsverfolgungKontakt
Biopsy specimen processing, CellSafe™
CellSafe™ biopsy capsule
  720-2333
 :   CELLPATH
Biopsy specimen processing, CellSafe™
  720-2333
 :  CELLPATH
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
CellSafe™ biopsy capsule
70,20 €
1 * 100 ST
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
  • Bezeichnung:
    CellSafe™ biopsy capsule