AuftragsverfolgungKontakt
Polystyrene, analytical standards for GPC
~275000 Mn, Mw/Mn ~1,03, 1 g
  81413-1G
 :   Merck
Polystyrene, analytical standards for GPC
  81413-1G
 :  Merck
 :  
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
~275000 Mn, Mw/Mn ~1,03, 1 g
 :  
191,00 €
1 * 1 g
 : Exact values can be taken from the accompanying certificate of analysis
 : Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
  • VE:
    1 g
  • Bezeichnung:
    ~275000 Mn, Mw/Mn ~1,03
  • Cat. No.:
    81413-1G
  • M-Clarity Qualitätsstufe:
    MQ100