Polystyrene, analytical standards for GPC
~275000 Mn, Mw/Mn ~1,03, 1 g
81413-1G
: Merck
: Exact values can be taken from the accompanying certificate of analysis
: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
- VE:1 g
- Bezeichnung:~275000 Mn, Mw/Mn ~1,03
- Cat. No.:81413-1G
- M-Clarity Qualitätsstufe:MQ100