Polystyrene, analytical standards for GPC
~187000 Mn, Mw/Mn ~1,05, 1 g
Artikelnr. 81412-1G
Anbieter: Merck
Springe zu:
Product Details & Documents
: Exact values can be taken from the accompanying certificate of analysis
: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
Spezifikationen
Spezifikationen
- VE:1 g
- Bezeichnung:~187000 Mn, Mw/Mn ~1,05
- Cat. No.:81412-1G
- M-Clarity Qualitätsstufe:MQ100
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.