Polystyrene, analytical standards for GPC
~89300 Mn, Mw/Mn ~1,06, 1 g
81410-1G
: Merck
Certifications: Exact values can be taken from the accompanying certificate of analysis
Caution: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
- VE:1 g
- Bezeichnung:~89300 Mn, Mw/Mn ~1,06
- Cat. No.:81410-1G
- M-Clarity Qualitätsstufe:MQ100
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.