Polystyrene, analytical standards for GPC
~64000 Mn, Mw/Mn ~1,02, 1 g
Artikelnr. 81409-1G
Anbieter: Merck
Springe zu:
Product Details & Documents
: Exact values can be taken from the accompanying certificate of analysis
: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
Spezifikationen
Spezifikationen
- VE:1 g
- Bezeichnung:~64000 Mn, Mw/Mn ~1,02
- Cat. No.:81409-1G
- M-Clarity Qualitätsstufe:MQ100