Polystyrene, analytical standards for GPC
~31000 Mn, Mw/Mn ~1,01, 250 mg
Catalog # 81408-250MG
Supplier: Merck
Jump to:
Product Details & Documents
Certifications: Exact values can be taken from the accompanying certificate of analysis
Caution: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
Specifications
Specifications
- VE:250 mg
- Bezeichnung:~31000 Mn, Mw/Mn ~1,01
- Cat. No.:81408-250MG
- M-Clarity Qualitätsstufe:MQ100