Express Eingabe
Austria
AuftragsverfolgungKontakt
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
~1790 Mn, Mw/Mn ~1,06, 1 g
Artikelnr. 81403-1G
Anbieter:   Merck
Polystyrene, analytical standards for GPC
Artikelnr. 81403-1G
Anbieter:  Merck
CAS Nummer:  
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
~1790 Mn, Mw/Mn ~1,06, 1 g
CAS Nummer:  
Preis
191,00 €
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents
Certifications: Exact values can be taken from the accompanying certificate of analysis
Caution: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
Spezifikationen
Spezifikationen
  • VE:
    1 g
  • Bezeichnung:
    ~1790 Mn, Mw/Mn ~1,06
  • Cat. No.:
    81403-1G
  • M-Clarity Qualitätsstufe:
    MQ100
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.