AuftragsverfolgungKontakt
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
~1020 Mn, Mw/Mn ~1,10, 1 g
  81402-1G
 :   Merck
Polystyrene, analytical standards for GPC
  81402-1G
 :  Merck
 :  
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
~1020 Mn, Mw/Mn ~1,10, 1 g
 :  
182,00 €
1 * 1 g
Certifications: Exact values can be taken from the accompanying certificate of analysis
Caution: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
  • VE:
    1 g
  • Bezeichnung:
    ~1020 Mn, Mw/Mn ~1,10
  • Cat. No.:
    81402-1G
  • M-Clarity Qualitätsstufe:
    MQ100