Express Eingabe
Austria
AuftragsverfolgungKontakt
n-Hexan, EMPLURA®, Supelco®

n-Hexan, EMPLURA®, Supelco®

Kunststoffflasche, 2,5 L
Catalog # 1.04368.2511
Flammable
Health
Irritant
Environment
Supplier:   Merck
n-Hexan, EMPLURA®, Supelco®
Catalog # 1.04368.2511
Supplier:  Merck
CAS Number:  110-54-3
n-Hexan, EMPLURA®, Supelco®
n-Hexan, EMPLURA®, Supelco®

Kunststoffflasche, 2,5 L

CAS Number:  110-54-3
Price
129,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Raumtemperatur
  • UN:1208
  • ADR:3,II
  •  :M-Clarity™ Qualitätsstufe = MQ200
  • EINECS:203-777-6

Documents

Specifications

Specifications

  • Packung:
    Kunststoffflasche
  • VE:
    2,5 L

Specifications