Express Eingabe
Austria
AuftragsverfolgungKontakt
Ubidecarenone, Reference Standards | Avantor

Ubidecarenone, Reference Standards

Ubidecarenone Related Compound A, USP reference standard, Ampule, 15 mg
Artikelnr. 1705323.
Anbieter:   USP
Ubidecarenone, Reference Standards
Artikelnr. 1705323.
Anbieter:  USP
CAS Nummer:  
Ubidecarenone, Reference Standards

Ubidecarenone Related Compound A, USP reference standard, Ampule, 15 mg

CAS Nummer:  
Preis
1 070,00 €
Einloggen für Verfügbarkeit
1 * 15 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Zertifizierung: US Pharmacopea Reference Standard

Achtung! Bitte beachten: Cold Shipment required

Spezifikationen

Spezifikationen

  • Packung:
    Ampule
  • VE:
    15 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Ubidecarenone Related Compound A
  • Cat. No.:
    1705323.
  • CAS Nr.:
    303-97-9
  • Synonym(s):
    Coenzyme Q9
  • Bezeichnung:
    Ubidecarenone Related Compound A

Spezifikationen