Cyclohexan, extra trocken 99.5%, AcroSeal®
Glass bottle, AcroSeal®, 100 mL
Anbieter: Thermo Fisher Scientific
Cyclohexan, extra trocken 99.5%, AcroSeal®
Artikelnr. 326831000.
Anbieter: Thermo Fisher Scientific
CAS Nummer: 110-82-7
Springe zu:
Product Details & Documents
Synonyme: Hexahydrobenzene, Hexamethylene
- CAS Nummer:110-82-7
- Formel:C₆H₁₂
- MDL-Nummer:MFCD00003814
- Molekulargewicht:84.16 g/mol
- Merck-Index:13,02752
- Siedepunkt:81 °C (760 mmHg)
- Schmelzpunkt:6,5 °C
- Dichte:0,77 g/cm³
- Blitzpunkt:–18 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Dokumente
ACRO32683
Sicherheitsdatenblatt (SDS)
Spezifikationen
Spezifikation Testergebnisse
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.001 meq/g
Spezifikationen
- Packung:Glass bottle, AcroSeal®
- VE:100 mL
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



