Express Eingabe
Austria
AuftragsverfolgungKontakt
Transport swabs
Amies agar Gel with Charcoal
Artikelnr. OXOITS0002A
Transport swabs
Artikelnr. OXOITS0002A
Transport swabs
Amies agar Gel with Charcoal
Preis
377,00 €
Einloggen für Verfügbarkeit
1 * 500 ST
Product Details & Documents
Effective transportation of throat, vaginal, wound and skin swabs conforming to highest medical device classification - Class II a surgically invasive transient use. All swabs have soft rayon tips, which are inert and non-toxic to micro-organisms.
The addition of charcoal in the medium is considered to neutralise bacterial toxins and other inhibitory substances and has been shown to increase the recovery of Neisseria gonorrhoeae.
  • Prevents disintegration of gel column during transport
  • Eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria
 : *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Spezifikationen
  • Bezeichnung:
    Amies agar Gel with Charcoal
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.