AuftragsverfolgungKontakt
Biotin conjugation estimation, HOOK™ BiotinQuant
HABA dye, 1 g
  BKC-03
Biotin conjugation estimation, HOOK™ BiotinQuant
  BKC-03
 :  
Biotin conjugation estimation, HOOK™ BiotinQuant
Biotin conjugation estimation, HOOK™ BiotinQuant
HABA dye, 1 g
 :  
148,00 €
1 * 1 g
HOOK™ BiotinQuant is designed for the estimation of biotin conjugation. It measures biotin using HABA [4’-hydroxyazobenzene-2-carboxylic acid] dye. HABA binds with avidin at the biotin-binding site.
  • VE:
    1 g
  • Bezeichnung:
    HABA dye