Express Eingabe
Austria
AuftragsverfolgungKontakt
 

n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

200 L
Catalog # 8044.9200
Flammable
Health
Irritant
Environment
Supplier:   Avantor
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Catalog # 8044.9200
Supplier:  Avantor
CAS Number:  110-54-3
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

200 L

CAS Number:  110-54-3
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specification Test Results

  • Assay (by GC):
    min. 95 %
  • Aromatic Compounds (as C6H6):
    max. 0,02 %
  • Color (APHA):
    max. 10
  • Density (g/ml) at 25°C:
    max. 0,657
  • Residue after Evaporation:
    max. 0,001 %
  • Sulfur Compounds (as S):
    max. 0,005 %
  • Thiophene:
    passes test
  • Water Soluble Titrable Acid (meq/g):
    max. 0,0003
  • Trace Impurities (in ppm):
    -
  • Aluminium (Al):
    max. 0,5
  • Barium (Ba):
    max. 0,1
  • Boron (B):
    max. 0,02
  • Cadmium (Cd):
    max. 0,05
  • Calcium (Ca):
    max. 0,5
  • Chromium (Cr):
    max. 0,02
  • Cobalt (Co):
    max. 0,02
  • Copper (Cu):
    max. 0,02
  • Iron (Fe):
    max. 0,1
  • Lead (Pb):
    max. 0,1
  • Magnesium (Mg):
    max. 0,1
  • Manganese (Mn):
    max. 0,02
  • Nickel (Ni):
    max. 0,02
  • Tin (Sn):
    max. 0,1
  • Zinc (Zn):
    max. 0,1

Specifications

  • VE:
    200 L

Specifications