Dosen mit Deckel
102 mm, 80 mm
Artikelnr. BOCH8620
Anbieter: BOCHEM
Springe zu:
Product Details & Documents
Jar made of stainless steel.
- Stackable
- Lid with handle
- Very good chemical and temperature resistance
Spezifikationen
- Ext.Ø:102 mm
- Höhe:80 mm