Büchner-Trichter
60 mm, 125 ml, 4
SMIT34S/60/4
Funnels, made of, borosilicate glass, with sintered glass disc. For convenient filtration of solutions.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Excellent temperature resistance
- Platte Ø:60 mm
- Porosität:4
- Kapazität:125 ml
- Seq:09



