Büchner-Trichter
40 mm, 75 ml, 4
Artikelnr. SMIT34S/40/4
Anbieter: SMITH SCIENTIFIC
Springe zu:
Product Details & Documents
Funnels, made of, borosilicate glass, with sintered glass disc. For convenient filtration of solutions.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Excellent temperature resistance
Spezifikationen
- Platte Ø:40 mm
- Porosität:4
- Kapazität:75 ml
- Seq:08
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



