Transmitted light
Transmitted light stage, with illuminated area 50 mm and int. Ø 84 mm
Artikelnr. 631-0885
Anbieter: SCHOTT AG Lighting and imaging
Springe zu:
Product Details & Documents
Dokumente
AT-ce_easyled_tls easyled_backlights_int_0914 TDS System Diagram for EasyLED Series TDS Transmitted Light Stage TDS Trasmitted Light Stage EasyLED and VisiLED
Technisches Datenblatt
Mehr
Spezifikationen
- Bezeichnung:Transmitted light stage, with illuminated area 50 mm and int. Ø 84 mm
- Seq:02