Express Eingabe
Austria
AuftragsverfolgungKontakt
Natriumcarbonat ≥97%, TECHNICAL

Natriumcarbonat ≥97%, TECHNICAL

Kunststoffeimer, 25 kg
Catalog # 27754.460
Irritant
Supplier:   VWR Chemicals
Natriumcarbonat ≥97%, TECHNICAL
Catalog # 27754.460
Supplier:  VWR Chemicals
CAS Number:  497-19-8
Natriumcarbonat ≥97%, TECHNICAL
Natriumcarbonat ≥97%, TECHNICAL

Kunststoffeimer, 25 kg

CAS Number:  497-19-8
Price
665,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Calcined soda, Carbonic acid disodium salt
  • CAS Number:497-19-8
  • Formula:Na₂CO₃
  • MDL Number:MFCD00003494
  • Molecular Weight:105.99 g/mol
  • Merck Index:14,08596
  • Boiling Pt:1600 °C (1013 hPa)
  • Melting Pt:854 °C
  • Density:2,53 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:207-838-8
  • REACH:01-2119485498-19
Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 97 %

Specifications

  • Packung:
    Kunststoffeimer
  • VE:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.