Express Eingabe
Austria
AuftragsverfolgungKontakt
Biological indicators, strips | Avantor
Biological indicators, strips
STRIP CONTROL GST E8/E6/E5/E4/E3 contain Geobacillus stearothermophilus (ATCC 7953) spores in predefined concentrations: E8=1-5x108CFU/strip; E6=1-5x106CFU/strip; E5=1-5x105CFU/strip; E4=1-5x104CFU/strip; E3=1-5x103CFU/strip.
Artikelnr. LIOF91055
Anbieter:   LIOFILCHEM
Biological indicators, strips
Artikelnr. LIOF91055
Anbieter:  LIOFILCHEM
Biological indicators, strips
STRIP CONTROL GST E8/E6/E5/E4/E3 contain Geobacillus stearothermophilus (ATCC 7953) spores in predefined concentrations: E8=1-5x108CFU/strip; E6=1-5x106CFU/strip; E5=1-5x105CFU/strip; E4=1-5x104CFU/strip; E3=1-5x103CFU/strip.
Preis
190,00 €
Einloggen für Verfügbarkeit
1 * 20 KIT
Product Details & Documents
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Spezifikationen
  • Bezeichnung:
    STRIP CONTROL GST E8/E6/E5/E4/E3 contain Geobacillus stearothermophilus (ATCC 7953) spores in predefined concentrations: E8=1-5x108CFU/strip; E6=1-5x106CFU/strip; E5=1-5x105CFU/strip; E4=1-5x104CFU/strip; E3=1-5x103CFU/strip.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.