Biological indicators, strips
STRIP CONTROL GST E8/E6/E5/E4/E3 contain Geobacillus stearothermophilus (ATCC 7953) spores in predefined concentrations: E8=1-5x108CFU/strip; E6=1-5x106CFU/strip; E5=1-5x105CFU/strip; E4=1-5x104CFU/strip; E3=1-5x103CFU/strip.
Anbieter: LIOFILCHEM
Springe zu:
Product Details & Documents
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Dokumente
Spezifikationen
- Bezeichnung:STRIP CONTROL GST E8/E6/E5/E4/E3 contain Geobacillus stearothermophilus (ATCC 7953) spores in predefined concentrations: E8=1-5x108CFU/strip; E6=1-5x106CFU/strip; E5=1-5x105CFU/strip; E4=1-5x104CFU/strip; E3=1-5x103CFU/strip.