Büchner-Trichter
60 mm, 125 ml, 3
Artikelnr. SMIT34S/60/3
Anbieter: SMITH SCIENTIFIC
Springe zu:
Product Details & Documents
Funnels, made of, borosilicate glass, with sintered glass disc. For convenient filtration of solutions.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Excellent temperature resistance
Spezifikationen
- Platte Ø:60 mm
- Porosität:3
- Kapazität:125 ml
- Seq:03



