Express Eingabe
Austria
AuftragsverfolgungKontakt
Glass wool
Glass wool
Deactivated glass wool, more inert than traditional glass wool, for vaporising a sample in a liner prior to introduction into a capillary column
Catalog # REST24324
Supplier:   Restek
Glass wool
Catalog # REST24324
Supplier:  Restek
Glass wool
Glass wool
Deactivated glass wool, more inert than traditional glass wool, for vaporising a sample in a liner prior to introduction into a capillary column
Price
60,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Specifications
  • Bezeichnung:
    Deactivated glass wool, more inert than traditional glass wool, for vaporising a sample in a liner prior to introduction into a capillary column
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.