Dissecting needles
Dissecting awl, length of spike 28 mm, S
Artikelnr. MURRD215/31
Anbieter: MURRAY S
Springe zu:
Product Details & Documents
Stainless steel spike in milled aluminium handle.
Spezifikationen
- Form:S
- Bezeichnung:Dissecting awl, length of spike 28 mm