AuftragsverfolgungKontakt
Accessories for data loggers, Dickson® | Avantor
Accessories for data loggers, Dickson®
Software and serial cable for data loggers, DicksonWare™ s
Artikelnr. DICKA015
Anbieter:   DICKSON
Accessories for data loggers, Dickson®
Artikelnr. DICKA015
Anbieter:  DICKSON
Accessories for data loggers, Dickson®
Software and serial cable for data loggers, DicksonWare™ s
Preis
176,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Spezifikationen
  • Bezeichnung:
    Software and serial cable for data loggers, DicksonWare™ s