AuftragsverfolgungKontakt
Density Bottles, Adjusted | Avantor
Density Bottles, Adjusted
50* ml
Artikelnr. LENZ05630228
Anbieter:   Lenz Laborglas
Density Bottles, Adjusted
Artikelnr. LENZ05630228
Anbieter:  Lenz Laborglas
Density Bottles, Adjusted
Density Bottles, Adjusted
50* ml
Preis
81,90 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
  • The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
  • The top of stopper is ground and polished for ease of wiping clean
  • Stopper is size NS 10/19 with capillary
* DIN ISO 3507
Dokumente
Spezifikationen
  • Volumen (nom.):
    50* ml