Twisselmann-Extraktionsapparate
Nur Kopf, 150 ml, 45/40 NS
Artikelnr. LENZ05415143
Anbieter: Lenz Laborglas
Springe zu:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Spezifikationen
- Extraktorinhalt:150 ml
- Hülse:45/40 NS
- Typ:Nur Kopf