Screw caps for screw thread tubes
GL 45, , ,
Artikelnr. LENZ01320045
Anbieter: Lenz Laborglas
Springe zu:
Product Details & Documents
PBT,red, with PTFE coated silicone sealing (VMQ).
- For DIN GL threads
- Heat resistant up to 180 °C
- Autoclavable, sterilisable with hot air
Spezifikationen
- Gewinde:GL 45
- Seq:05