Hydroxylammoniumchlorid 99%
Glass bottle for solids, 1.000 g
Anbieter: Thermo Fisher Scientific
Hydroxylammoniumchlorid 99%
Artikelnr. A15398.0B
Anbieter: Thermo Fisher Scientific
CAS Nummer: 5470-11-1
Springe zu:
Product Details & Documents
Purity: 99% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Odour: Acidic pH: 3.6 Melting Point: 152° dec. Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089
Synonyme: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL-Nummer:MFCD00051089
- Molekulargewicht:69.49 g/mol
- Merck-Index:14,4828
- Siedepunkt:306 °C (1013 hPa)
- Schmelzpunkt:152 °C (Zers.)
- Schmelzpunkt:152 °C (Zersetzung)
- Dichte:1,67
- Lagertemperatur:Raumtemperatur
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Documents
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:1.000 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



