Express Eingabe
Austria
AuftragsverfolgungKontakt
Adipohydrazid ≥97%

Adipohydrazid ≥97%

Glasflasche für Feststoffe, 100 g
Catalog # A15119.22
Irritant
Adipohydrazid ≥97%
Catalog # A15119.22
CAS Number:  1071-93-8
Adipohydrazid ≥97%
Adipohydrazid ≥97%

Glasflasche für Feststoffe, 100 g

CAS Number:  1071-93-8
Price
105,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 1071-93-8 Molecular Formula: H2NNHCO(CH2)4CONHNH2 Formula Weight: 174.20 Odour: Odourless Melting Point: 180-182° Beilstein Reference: 973863 MDL No.: MFCD00007614

Synonyms:  Adipyl hydrazide
  • CAS Number:1071-93-8
  • Formula:C₆H₁₄N₄O₂
  • MDL Number:MFCD00007614
  • Molecular Weight:174.2 g/mol
  • Boiling Pt:446 °C (1013 hPa)
  • Melting Pt:180…182 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:213-999-5
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.