Adenin ≥99%
Glass bottle for solids, 250 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 99% CAS No.: 73-24-5 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odour: Odourless Melting Point: 360° dec. Beilstein Reference: 5777 Merck Reference: 14,152 UN No.: 2811 MDL No.: MFCD00041790
Synonyme: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
- CAS Nummer:73-24-5
- Formel:C₅H₅N₅
- MDL-Nummer:MFCD00041790
- Molekulargewicht:135.13 g/mol
- Merck-Index:14,152
- Schmelzpunkt:>360 °C (Zers.)
- Schmelzpunkt:>360 °C (Zersetzung)
- Dichte:0,99741 g/cm³ (25 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:200-796-1
- UN:2811
- ADR:6.1,III
Documents
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:250 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



