n-Decan ≥99%
Glasflasche, 2.500 mL
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 99% CAS No.: 124-18-5 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.730 Refractive Index: 1.4115 Appearance: Colourless Odour: Petrol-like Melting Point: -30° Boiling Point: 172-174° Solubility: Soluble in alcohol and ethers Beilstein Reference: 1696981 UN No.: 2247 MDL No.: MFCD00008954
- CAS Nummer:124-18-5
- Formel:H₃C(CH₂)₈CH₃
- MDL-Nummer:MFCD00008954
- Molekulargewicht:142.28 g/mol
- Siedepunkt:172…174 °C
- Schmelzpunkt:–30 °C
- Dichte:0,730
- Blitzpunkt:46 °C (115 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:204-686-4
- UN:2247
- ADR:3,III
Spezifikationen
Spezifikationen
- Packung:Glasflasche
- VE:2.500 mL



