Harnsäure ≥99%
Glass bottle for solids, 100 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 99% CAS No.: 69-93-2 Molecular Formula: C5H4N4O3 Formula Weight: 168.11 Melting Point: 300° Beilstein Reference: 156158 Merck Reference: 14,9875 MDL No.: MFCD00005712
Synonyme: 1H-Purine-2,6,8(3H,7H,9H)-trione
- CAS Nummer:69-93-2
- Formel:C₅H₄N₄O₃
- MDL-Nummer:MFCD00005712
- Molekulargewicht:168.11 g/mol
- Merck-Index:14,9875
- Schmelzpunkt:>300 °C
- Dichte:1,89
- Lagertemperatur:Raumtemperatur
- EINECS:200-720-7
Documents
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



