Dibenzyldiselenid 95%
Glass bottle for solids, 25 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 95% CAS No.: 1482-82-2 Molecular Formula: C6H5CH2Se-SeCH2C6H5 Formula Weight: 340.18 Melting Point: 91-93° Beilstein Reference: 1877745 UN No.: 3283 MDL No.: MFCD00004767
- CAS Nummer:1482-82-2
- Formel:C₁₄H₁₄Se₂
- MDL-Nummer:MFCD00004767
- Molekulargewicht:340.19 g/mol
- Schmelzpunkt:91…93 °C
- Dichte:1,67 g/cm³ (25 °C)
- Lagertemperatur:Raumtemperatur
- UN:3283
- ADR:6.1,III
- :Dichte: 1,67 g/cm³ (25 °C)
Dokumente
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:25 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



