Express Eingabe
Austria
AuftragsverfolgungKontakt
Dibenzyldiselenid 95%

Dibenzyldiselenid 95%

Glass bottle for solids, 5 g
Catalog # B21278.06
Toxic
Health
Environment
Dibenzyldiselenid 95%
Catalog # B21278.06
CAS Number:  1482-82-2
Dibenzyldiselenid 95%
Dibenzyldiselenid 95%

Glass bottle for solids, 5 g

CAS Number:  1482-82-2
Price
114,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 95% CAS No.: 1482-82-2 Molecular Formula: C6H5CH2Se-SeCH2C6H5 Formula Weight: 340.18 Melting Point: 91-93° Beilstein Reference: 1877745 UN No.: 3283 MDL No.: MFCD00004767

  • CAS Number:1482-82-2
  • Formula:C₁₄H₁₄Se₂
  • MDL Number:MFCD00004767
  • Molecular Weight:340.19 g/mol
  • Melting Pt:91…93 °C
  • Density:1,67 g/cm³ (25 °C)
  • Storage Temperature:Raumtemperatur
  • UN:3283
  • ADR:6.1,III
  •  :Dichte: 1,67 g/cm³ (25 °C)
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    5 g

Specifications