Auftragserfassung
Austria
AuftragsverfolgungKontakt
D(+)-Kampfer ≥98%

D(+)-Kampfer ≥98%

Glasflasche für Feststoffe, 500 g
Catalog # A10708.36
Flammable
Irritant
D(+)-Kampfer ≥98%
Catalog # A10708.36
CAS Number:  464-49-3
D(+)-Kampfer ≥98%
D(+)-Kampfer ≥98%

Glasflasche für Feststoffe, 500 g

CAS Number:  464-49-3
Price
217,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 464-49-3 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 65°(149°F) Odour: Camphor-like Boiling Point: 204° Beilstein Reference: 2042745 Merck Reference: 14,1732 Optical Rotation: +43° (c=10 in ethanol) UN No.: 2717 MDL No.: MFCD00064149

Synonyms:  (R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (1R,4R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R,4R)-(+)-1,7,7-Trimethyl-2-norbornanone, (R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (+)-Camphor
  • CAS Number:464-49-3
  • Formula:C₁₀H₁₆O
  • MDL Number:MFCD00064149
  • Molecular Weight:152.24 g/mol
  • Merck Index:14,1732
  • Boiling Pt:204 °C
  • Melting Pt:175…180 °C
  • Density:0,990
  • Flash Pt:65 °C (149 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:207-355-2
  • UN:2717
  • ADR:4.1,III
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.