D(+)-Kampfer ≥98%
Glasflasche für Feststoffe, 100 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 98% CAS No.: 464-49-3 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 65°(149°F) Odour: Camphor-like Boiling Point: 204° Beilstein Reference: 2042745 Merck Reference: 14,1732 Optical Rotation: +43° (c=10 in ethanol) UN No.: 2717 MDL No.: MFCD00064149
Synonyme: (R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (1R,4R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R,4R)-(+)-1,7,7-Trimethyl-2-norbornanone, (R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (+)-Camphor
- CAS Nummer:464-49-3
- Formel:C₁₀H₁₆O
- MDL-Nummer:MFCD00064149
- Molekulargewicht:152.24 g/mol
- Merck-Index:14,1732
- Siedepunkt:204 °C
- Schmelzpunkt:175…180 °C
- Dichte:0,990
- Blitzpunkt:65 °C (149 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:207-355-2
- UN:2717
- ADR:4.1,III
Documents
Spezifikationen
Spezifikationen
- Packung:Glasflasche für Feststoffe
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



