4'-Hydroxy-3'-methoxyacetophenon ≥98%
Glass bottle for solids, 50 g
Anbieter: Thermo Fisher Scientific
4'-Hydroxy-3'-methoxyacetophenon ≥98%
Artikelnr. A10439.18
Anbieter: Thermo Fisher Scientific
CAS Nummer: 498-02-2
Springe zu:
Product Details & Documents
Purity: 98% CAS No.: 498-02-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 112-116° Boiling Point: 295-300° Merck Reference: 14,741 MDL No.: MFCD00008747
Synonyme: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
- CAS Nummer:498-02-2
- Formel:C₉H₁₀O₃
- MDL-Nummer:MFCD00008747
- Molekulargewicht:166.18 g/mol
- Merck-Index:14,741
- Siedepunkt:295…300 °C
- Schmelzpunkt:112…116 °C
- Lagertemperatur:Raumtemperatur
- EINECS:207-854-5
Documents
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:50 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



