Dimethyldisulfid ≥99%
Glasflasche für Feststoffe, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 624-92-0 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 10°(50°F) Refractive Index: 1.5253 Odour: Stench Melting Point: -85° Boiling Point: 108-109° Beilstein Reference: 1730824 UN No.: 2381 MDL No.: MFCD00008561
Synonyms: Dimethyl disulphide, Methyl disulphide
- CAS Number:624-92-0
- Formula:C₂H₆S₂
- MDL Number:MFCD00008561
- Molecular Weight:94.2 g/mol
- Boiling Pt:108…109 °C
- Melting Pt:–85 °C
- Density:1,046
- Flash Pt:10 °C (50 °F)
- Storage Temperature:Raumtemperatur
- EINECS:210-871-0
- UN:2381
- ADR:3,II
Documents
Certificates
Search by Lot NumberALFAL09679.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:1.000 g



