Slide mailers
Slide mailers, Weiß, PP, 2/4
Artikelnr. SARS86.972.150
Marketsource
Anbieter: SARSTEDT
Springe zu:
Product Details & Documents
These mailing vessels are manufactured from polypropylene and is suitable for the safe transportation or storage of two glass microscope slides (or four if placed against each other).
- The lid features a ventilation hole
Ordering information: Containers available with or without ventilation hole.
Packaging: 50 pieces per sachet.
Spezifikationen
- Anzahl Objektträger:2/4
- Farbe:Weiß
- Bezeichnung:Slide mailers
- Material:PP
- Seq:01