AuftragsverfolgungKontakt
Microtubes, Prepared | Avantor
Microtubes, Prepared
Microtubes, prepared, coagulation citrate, with attached push cap closure, 1,3 ml, 1000/case
Artikelnr. SARS41.1506.005
Marketsource
Anbieter:   SARSTEDT
Microtubes, Prepared
Artikelnr. SARS41.1506.005
Anbieter:  SARSTEDT
Marketsource
Microtubes, Prepared
Microtubes, prepared, coagulation citrate, with attached push cap closure, 1,3 ml, 1000/case
Preis
188,00 €
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Spezifikationen
  • Verpackt:
    1000/case
  • Kapazität:
    1,3 ml
  • Bezeichnung:
    Microtubes, prepared, coagulation citrate, with attached push cap closure
  • Seq:
    15