Staining baskets and racks
Staining basket, for 45 horizontal, 235×85 mm
Artikelnr. DELTCP-45H
Anbieter: DELTALAB
Springe zu:
Product Details & Documents
Staining baskets and racks, made of stainless steel. Designed for 76×26 mm slides.
- Good chemical resistance
- Good temperature resistance
Spezifikationen
- Bezeichnung:Staining basket, for 45 horizontal, 235×85 mm
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



