Xanthydrol ≥98%
Glass bottle for solids, 5 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Purity: 98+% CAS No.: 90-46-0 Molecular Formula: C13H10O2 Formula Weight: 198.22 Storage Temperature: 2°C to 8°C Melting Point: 122-126° Beilstein Reference: 10395 MDL No.: MFCD00005057
Synonyme: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
- CAS Nummer:90-46-0
- Formel:C₁₃H₁₀O₂
- MDL-Nummer:MFCD00005057
- Molekulargewicht:198.22 g/mol
- Siedepunkt:338 °C (1013 hPa)
- Schmelzpunkt:122…126 °C
- EINECS:201-996-1
Documents
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:5 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



