AuftragsverfolgungKontakt
Polyoxyl stearate, Reference Standards | Avantor
Polyoxyl stearate, Reference Standards
Polyoxyl 40 Stearate, USP reference standard, Vial, 200 mg
Artikelnr. 1547903.
Anbieter:   USP
Polyoxyl stearate, Reference Standards
Artikelnr. 1547903.
Anbieter:  USP
CAS Nummer:  
Polyoxyl stearate, Reference Standards
Polyoxyl 40 Stearate, USP reference standard, Vial, 200 mg
CAS Nummer:  
Preis
373,00 €
Einloggen für Verfügbarkeit
1 * 200 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zertifizierung: US Pharmacopea Reference Standard
Spezifikationen
Spezifikationen
  • Packung:
    Vial
  • VE:
    200 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Polyoxyl 40 Stearate
  • Cat. No.:
    1547903.
  • CAS Nr.:
    9004-99-3
  • Bezeichnung:
    Polyoxyl 40 Stearate
Spezifikationen